Methyl-3-amino-5-phenyl-2-thiophencarboxylat

Lieferant: FLUOROCHEM

Synonyme: Methyl 3-amino-5-phenylthiophene-2-carboxylate

CFF195.2.5 CFF195.10 CFF195.25 017264-25G 017264-5G
CHMPCFF195.2.5EA 268 EUR
CHMPCFF195.2.5 CHMPCFF195.10 CHMPCFF195.25 FLUO017264-25G FLUO017264-5G
Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Formel: C₁₂H₁₁NO₂S
Molecular Weight: 233.2862 g/mol
MDL: MFCD00068161
CAS-Nummer: 100063-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR