Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Lieferant: Thermo Scientific
Synonyme:
Methyl 3-amino-5-phenylthiophene-2-carboxylate
GK00498DA
GK00498EA
MAYBGK00498DAEA
0
EUR
MAYBGK00498DA
MAYBGK00498EA
Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Formel:
C₁₂H₁₁NO₂S Molecular Weight: 233,2862 g/mol |
MDL:
MFCD00068161 CAS-Nummer: 100063-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...