(S)-(-)-2-Pyrrolidinon-5-carbonsäure 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
L-Pyroglutamic Acid, (S)-(-)-2-Pyrrolidone-5-carboxylic acid, H-Pyr-OH, (S)-5-Oxopyrrolidine-2-carboxylic acid
132121000
132120250
132125000
ACRO132121000EA
59.5
EUR
ACRO132121000
ACRO132120250
ACRO132125000
(S)-(-)-2-Pyrrolidinon-5-carbonsäure 98%
(S)-(-)-2-Pyrrolidinon-5-carbonsäure
Formel:
C₅H₇NO₃ Molecular Weight: 129,1 g/mol Schmelzpunkt: 152…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005272 CAS-Nummer: 98-79-3 EINECS: 202-700-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Specific optical rotation | -26° to -29° (20°C, 589 nm) (c=10, 1N NaOH) (on dry substance) |
Titration with NaOH | ≥97.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...