(S)-(-)-2-Pyrrolidinon-5-carbonsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
L-Pyroglutamic Acid, (S)-(-)-2-Pyrrolidone-5-carboxylic acid, H-Pyr-OH, (S)-5-Oxopyrrolidine-2-carboxylic acid
L11711.14
L11711.22
L11711.36
L11711.14EA
22.2
EUR
L11711.14
L11711.22
L11711.36
(S)-(-)-2-Pyrrolidinon-5-carbonsäure ≥98%
(S)-(-)-2-Pyrrolidinon-5-carbonsäure
Purity: 98+%
CAS No.: 98-79-3
Molecular Formula: C5H7NO3
Formula Weight: 129.12
Melting Point: 158-162°
Beilstein Reference: 82132
Merck Reference: 14,8001
Optical Rotation: -10.5° (c=5 in water)
MDL No.: MFCD00005272
Formel:
C₅H₇NO₃ Schmelzpunkt: 158…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005272 CAS-Nummer: 98-79-3 EINECS: 202-700-3 Merck Index: 14,8001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...