(S)-(-)-2-Pyrrolidinon-5-carbonsäure

Lieferant: BACHEM BIOCHEMICA

Synonyme: L-Pyroglutamic Acid, (S)-(-)-2-Pyrrolidone-5-carboxylic acid, H-Pyr-OH, (S)-5-Oxopyrrolidine-2-carboxylic acid

4026127.0100 4026127.0500
BACE4026127.0100EA 0 EUR
BACE4026127.0100 BACE4026127.0500
(S)-(-)-2-Pyrrolidinon-5-carbonsäure
(S)-(-)-2-Pyrrolidinon-5-carbonsäure
Formel: C₅H₇NO₃
Molecular Weight: 129,1 g/mol
MDL: MFCD00005272
CAS-Nummer: 98-79-3
EINECS: 202-700-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR