3-Ethyl-3-oxetanmethanol ≥97%
Lieferant: Thermo Scientific
Synonyme:
(3-Ethyloxetan-3-yl)methanol, 2-Hydroxymethyl-2-ethyl-1,3-epoxypropane, 3-Hydroxymethyl-3-ethyloxetane
H53497.06
H53497.14
H53497.06EA
35.1
EUR
H53497.06
H53497.14
3-Ethyl-3-oxetanmethanol ≥97%
3-Ethyl-3-oxetanmethanol
Purity: 97%
CAS No.: 3047-32-3
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 108°(226°F)
Refractive Index: 1.453
Boiling Point: 96°/4mm
MDL No.: MFCD00190143
Formel:
C₆H₁₂O₂ Molecular Weight: 116,16 g/mol Siedepunkt: 96 °C (4 mmHg) Dichte: 1,019 Flammpunkt: 108 °C (226 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190143 CAS-Nummer: 3047-32-3 EINECS: 221-254-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...