Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl)

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl

4002258.0005 4002258.0025
BACE4002258.0005EA 0 EUR
BACE4002258.0005 BACE4002258.0025
Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl)
Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl)
Formel: C₆H₁₄ClNO₂
Molecular Weight: 167.64 g/mol
MDL: MFCD00058255
CAS-Nummer: 27532-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR