Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl) 98%

Lieferant: Thermo Scientific

Synonyme: Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl

120101000 120105000
ACRO120101000EA 31.2 EUR
ACRO120101000 ACRO120105000
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl) 98%
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)
Formel: C₃H₈ClNO₂
Molecular Weight: 125,56 g/mol
Schmelzpunkt: 173…177 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012870
CAS-Nummer: 5680-79-5
EINECS: 227-139-1

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %
GC ≥95 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR