Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl
4030678.0250
4030678.1000
BACE4030678.0250EA
0
EUR
BACE4030678.0250
BACE4030678.1000
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)
Formel:
C₃H₈ClNO₂ Molecular Weight: 125,56 g/mol |
MDL:
MFCD00012870 CAS-Nummer: 5680-79-5 EINECS: 227-139-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...