Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl

M-Clarity™ Qualitätsstufe = MQ200

G6600-100G G6600-500G G6600-5G
SIALG6600-100GEA 58.5 EUR
SIALG6600-100G SIALG6600-500G SIALG6600-5G
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl), Sigma-Aldrich®
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)
Formel: C₃H₈ClNO₂
Molecular Weight: 125,56 g/mol
MDL: MFCD00012870
CAS-Nummer: 5680-79-5
EINECS: 227-139-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR