Triethyl-1,1,2-ethantricarboxylat
Lieferant: FLUOROCHEM
024178-100G
024178-25G
FLUO024178-100GEA
195
EUR
FLUO024178-100G
FLUO024178-25G
Triethyl-1,1,2-ethantricarboxylat
Triethyl-1,1,2-ethantricarboxylat
Formel:
C₁₁H₁₈O₆ |
MDL:
MFCD00009154 CAS-Nummer: 7459-46-3 EINECS: 231-235-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...