Triethyl-1,1,2-ethantricarboxylat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Triethyl ethane-1,1,2-tricarboxylate
M-Clarity™ Qualitätsstufe = MQ100
T59854-100G
T59854-25G
SIALT59854-100GEA
190
EUR
SIALT59854-100G
SIALT59854-25G
Triethyl-1,1,2-ethantricarboxylat, Sigma-Aldrich®
Triethyl-1,1,2-ethantricarboxylat
Formel:
C₁₁H₁₈O₆ Molecular Weight: 246,26 g/mol |
MDL:
MFCD00009154 CAS-Nummer: 7459-46-3 EINECS: 231-235-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...