Phenylcarbamat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
P21404-25G
P21404-5G
SIALP21404-25GEA
190
EUR
SIALP21404-25G
SIALP21404-5G
Phenylcarbamat, Sigma-Aldrich®
Phenylcarbamat
Formel:
C₇H₇NO₂ Molecular Weight: 137,14 g/mol |
MDL:
MFCD00007961 CAS-Nummer: 622-46-8 EINECS: 210-737-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...