Trimethylthioharnstoff

Lieferant: Thermo Scientific

NH00387.1 NH00387.10 NH00387.50
MAYBNH00387.1EA 0 EUR
MAYBNH00387.1 MAYBNH00387.10 MAYBNH00387.50
Trimethylthioharnstoff
Trimethylthioharnstoff
Formel: C₄H₁₀N₂S
MDL: MFCD00059148
CAS-Nummer: 2489-77-2
EINECS: 219-644-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR