2-Aminoethylhydrogensulfat 99%

Lieferant: Thermo Scientific

Synonyme: Sulfuric acid Mono(2-aminoethyl) Ester, 2-Aminoethyl Hydrogen sulphate

103641000 103645000
ACRO103641000EA 76.9 EUR
ACRO103641000 ACRO103645000
2-Aminoethylhydrogensulfat 99%
2-Aminoethylhydrogensulfat
Formel: C₂H₇NO₄S
Molecular Weight: 141,1500 g/mol
Schmelzpunkt: 277…280 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008179
CAS-Nummer: 926-39-6
EINECS: 213-135-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR