2-Aminoethylhydrogensulfat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Sulfuric acid Mono(2-aminoethyl) Ester, 2-Aminoethyl Hydrogen sulphate
M-Clarity™ Qualitätsstufe = MQ100
06720-100G
06720-500G
SIAL06720-100GEA
59.6
EUR
SIAL06720-100G
SIAL06720-500G
2-Aminoethylhydrogensulfat, Sigma-Aldrich®
2-Aminoethylhydrogensulfat
Formel:
C₂H₇NO₄S Molecular Weight: 141,1500 g/mol |
MDL:
MFCD00008179 CAS-Nummer: 926-39-6 EINECS: 213-135-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...