(Methylamino)acetaldehyddimethylacetal 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,2-Dimethoxy-N-methylethanamine
L02391.14
L02391.22
L02391.14EA
39.7
EUR
L02391.14
L02391.22
(Methylamino)acetaldehyddimethylacetal 97%
(Methylamino)acetaldehyddimethylacetal
Purity: 97%
CAS No.: 122-07-6
Molecular Formula: CH3NHCH2CH(OCH3)2
Formula Weight: 119.16
Flash Point: 29°(84°F)
Refractive Index: 1.4115
Odour: Ammonia-like
pH: 10.9
Melting Point: -73°
Boiling Point: 139-140°
Beilstein Reference: 605322
UN No.: 1993
MDL No.: MFCD00008485
Formel:
C₅H₁₃NO₂ Molecular Weight: 119,16 g/mol Siedepunkt: 139…140 °C Schmelzpunkt: –73 °C Dichte: 0,928 Flammpunkt: 29 °C (84 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008485 CAS-Nummer: 122-07-6 EINECS: 204-520-0 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...