N-Benzylglycinethylester ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: Ethyl 2-(benzylamino)acetate

L15496.14 L15496.22 L15496.36
L15496.14EA 49.4 EUR
L15496.14 L15496.22 L15496.36
N-Benzylglycinethylester ≥97%
N-Benzylglycinethylester

Purity: 97% CAS No.: 6436-90-4 Molecular Formula: C6H5CH2NHCH2CO2CH2CH3 Formula Weight: 193.25 Storage Temperature: 2°C to 8°C Flash Point: 112°(233°F) Refractive Index: 1.5053 Boiling Point: 102-104°/1mm Beilstein Reference: 2104816 MDL No.: MFCD00009174

Formel: C₁₁H₁₅NO₂
Molecular Weight: 193.25 g/mol
Siedepunkt: 102…104 °C (1 mmHg)
Dichte: 1.048
Flammpunkt: 112 °C (233 °F)
MDL: MFCD00009174
CAS-Nummer: 6436-90-4
EINECS: 229-218-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR