N-Benzylglycinethylester

Lieferant: CHEMPUR

Synonyme: Ethyl 2-(benzylamino)acetate

FL11445.POR 011445-100G
CHMPFL11445.POREA 0 EUR
CHMPFL11445.POR FLUO011445-100G
N-Benzylglycinethylester
N-Benzylglycinethylester
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193.25 g/mol
MDL: MFCD00009174
CAS-Nummer: 6436-90-4
EINECS: 229-218-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR