3-(Trimethoxysilyl)propylmethacrylat, Millipore®
Lieferant: Merck
Synonyme:
γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane
M6514-25ML
M6514-50ML
SIALM6514-25MLEA
90.7
EUR
SIALM6514-25ML
SIALM6514-50ML
3-(Trimethoxysilyl)propylmethacrylat, Millipore®
3-(Trimethoxysilyl)propylmethacrylat
Formel:
C₁₀H₂₀O₅Si Molecular Weight: 248,35 g/mol |
MDL:
MFCD00008593 CAS-Nummer: 2530-85-0 EINECS: 219-785-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...