3-(Trimethoxysilyl)propylmethacrylat

Lieferant: CHEMPUR SILANE

SILS10494.25 SILS10494.100
SILNSILS10494.25EA 0 EUR
SILNSILS10494.25 SILNSILS10494.100
3-(Trimethoxysilyl)propylmethacrylat
3-(Trimethoxysilyl)propylmethacrylat
Formel: C₁₀H₂₀O₅Si
MDL: MFCD00008593
CAS-Nummer: 2530-85-0
EINECS: 219-785-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR