3-(Trimethoxysilyl)propylmethacrylat
Lieferant: CHEMPUR SILANE
Synonyme:
γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane
SILS10750.50
SILS10750.100
SILS10750.500
SILNSILS10750.50EA
0
EUR
SILNSILS10750.50
SILNSILS10750.100
SILNSILS10750.500
3-(Trimethoxysilyl)propylmethacrylat
3-(Trimethoxysilyl)propylmethacrylat
Formel:
C₁₀H₂₀O₅Si Molecular Weight: 248.35 g/mol |
MDL:
MFCD00008593 CAS-Nummer: 2530-85-0 EINECS: 219-785-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...