3-Bromdiphenylether (BDE Nr. 2)

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromodiphenyl ether, 1-Bromo-3-phenoxybenzene

OT-0818-5G OT-0818-1G
COBBOT-0818-5GEA 262 EUR
COBBOT-0818-5G COBBOT-0818-1G
3-Bromdiphenylether (BDE Nr. 2)
3-Bromdiphenylether (BDE Nr. 2)
Formel: C₁₂H₉BrO
Molecular Weight: 249,1000 g/mol
MDL: MFCD00474520
CAS-Nummer: 6876-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR