Kupferron, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ammonium 2-oxo-1-phenylhydrazinolate

675636-100G 675636-500G
SIAL675636-100GEA 322 EUR
SIAL675636-100G SIAL675636-500G
Kupferron, Sigma-Aldrich®
Kupferron
Formel: C₆H₉N₃O₂
Molecular Weight: 155,16 g/mol
MDL: MFCD00078422
CAS-Nummer: 135-20-6
EINECS: 205-183-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR