3-Phenoxyanisol 97

Lieferant: Thermo Fisher Scientific

H32482.06 H32482.14
H32482.06EA 51.3 EUR
H32482.06 H32482.14
3-Phenoxyanisol 97
3-Phenoxyanisol

Purity: 97% CAS No.: 1655-68-1 Molecular Formula: C13H12O2 Formula Weight: 200.23 Refractive Index: 1.5790 Odour: Odourless MDL No.: MFCD00455605

Formel: C₁₃H₁₂O₂
Molecular Weight: 200.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00455605
CAS-Nummer: 1655-68-1
EINECS: 216-748-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR