2-(Methoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid
BB-2125-1G
BB-2125-5G
BB-2125-25G
COBBBB-2125-1GEA
35.9
EUR
COBBBB-2125-1G
COBBBB-2125-5G
COBBBB-2125-25G
2-(Methoxycarbonyl)phenylboronsäure
2-(Methoxycarbonyl)phenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179,97 g/mol |
MDL:
MFCD02179452 CAS-Nummer: 374538-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...