3-(Methoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 3-boronobenzoate, 3-(Methoxycarbonyl)benzeneboronic acid
BB-2116-5G
BB-2116-25G
BB-2116-1G
BB-2116-100G
COBBBB-2116-5GEA
0
EUR
COBBBB-2116-5G
COBBBB-2116-25G
COBBBB-2116-1G
COBBBB-2116-100G
3-(Methoxycarbonyl)phenylboronsäure
3-(Methoxycarbonyl)phenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179.97 g/mol |
MDL:
MFCD02093046 CAS-Nummer: 99769-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...