3-(Methoxycarbonyl)phenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
Methyl 3-boronobenzoate, 3-(Methoxycarbonyl)benzeneboronic acid
010804-5G
010804-25G
FLUO010804-5GEA
60.9
EUR
FLUO010804-5G
FLUO010804-25G
3-(Methoxycarbonyl)phenylboronsäure
3-(Methoxycarbonyl)phenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179.97 g/mol |
MDL:
MFCD02093046 CAS-Nummer: 99769-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...