4-(Methoxycarbonyl)phenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid
H27627.03
H27627.06
H27627.03EA
71.2
EUR
H27627.03
H27627.06
4-(Methoxycarbonyl)phenylboronsäure ≥97%
4-(Methoxycarbonyl)phenylboronsäure
Purity: 97%
CAS No.: 99768-12-4
Molecular Formula: C8H9BO4
Formula Weight: 179.97
Melting Point: 220-225°
MDL No.: MFCD01632203
Formel:
C₈H₉BO₄ Molecular Weight: 179,97 g/mol Schmelzpunkt: 220…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01632203 CAS-Nummer: 99768-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...