4-(Methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2462-5G BB-2462-25G BB-2462-100G
COBBBB-2462-5GEA 0 EUR
COBBBB-2462-5G COBBBB-2462-25G COBBBB-2462-100G
4-(Methoxycarbonyl)phenylboronsäure
4-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
MDL: MFCD01632203
CAS-Nummer: 99768-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR