4-(Methoxycarbonyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

594539-1G 594539-5G
SIAL594539-1GEA 43.3 EUR
SIAL594539-1G SIAL594539-5G
4-(Methoxycarbonyl)phenylboronsäure, Sigma-Aldrich®
4-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179,97 g/mol
MDL: MFCD01632203
CAS-Nummer: 99768-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR