4-Benzyloxyphenylacetonitril
Lieferant: FLUOROCHEM
Synonyme:
2-(4-(Benzyloxy)phenyl)acetonitrile
018228-1G
018228-5G
FLUO018228-1GEA
34.1
EUR
FLUO018228-1G
FLUO018228-5G
4-Benzyloxyphenylacetonitril
4-Benzyloxyphenylacetonitril
Formel:
C₁₅H₁₃NO Molecular Weight: 223.28 g/mol |
MDL:
MFCD00016400 CAS-Nummer: 838-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...