(2S)-2-Pyrrolidinylmethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol

M-Clarity™ Qualitätsstufe = MQ100

186511-25G 186511-1G 186511-5G
SIAL186511-25GEA 60.4 EUR
SIAL186511-25G SIAL186511-1G SIAL186511-5G
(2S)-2-Pyrrolidinylmethanol, Sigma-Aldrich®
(S)-(+)-Prolinol
Formel: C₅H₁₁NO
Molecular Weight: 101,15 g/mol
MDL: MFCD00005255
CAS-Nummer: 23356-96-9
EINECS: 245-605-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR