4-(Benzyloxy)-3-methoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

163619-100G 163619-25G
SIAL163619-100GEA 105 EUR
SIAL163619-100G SIAL163619-25G
4-(Benzyloxy)-3-methoxybenzaldehyd, Sigma-Aldrich®
4-(Benzyloxy)-3-methoxybenzaldehyd
Formel: C₁₅H₁₄O₃
Molecular Weight: 242,27 g/mol
MDL: MFCD00003365
CAS-Nummer: 2426-87-1
EINECS: 219-379-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR