N,N,N',N'-Tetramethylbenzidin 97.5%

Lieferant: Thermo Fisher Scientific

Synonyme: N,n,n,n-tetramethylbenzidine, 4,4'-Bis(dimethylamino)biphenyl

138380050 138380250
ACRO138380050EA 96.1 EUR
ACRO138380050 ACRO138380250
N,N,N',N'-Tetramethylbenzidin 97.5%
N,N,N',N'-Tetramethylbenzidin
Formel: C₁₆H₂₀N₂
Schmelzpunkt: 192…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008310
CAS-Nummer: 366-29-0
EINECS: 206-676-5

Bestellen

Spezifikation Testergebnisse

Appearance Brown Powder
Infrared spectrum Conforms
Melting point 192 to 196 °C
Titration with HClO4 ≥96.5 %
HPLC ≥97.5 %
Specific optical rotation +76° to +83° (20 °C, 589 nm) (c=1, dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR