6-Methoxyisochinolin

Lieferant: COMBI-BLOCKS

HC-6022-5G HC-6022-1G
COBBHC-6022-5GEA 0 EUR
COBBHC-6022-5G COBBHC-6022-1G
6-Methoxyisochinolin
6-Methoxyisoquinoline
Formel: C₁₀H₉NO
MDL: MFCD04114861
CAS-Nummer: 52986-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR