6-Methoxyisochinolin

Lieferant: CHEMPUR

50-0054.1 50-0054.5
CHMP50-0054.1EA 0 EUR
CHMP50-0054.1 CHMP50-0054.5
6-Methoxyisochinolin
6-Methoxyisoquinoline
Formel: C₁₀H₉NO
Molecular Weight: 159,1846 g/mol
MDL: MFCD04114861
CAS-Nummer: 52986-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR