7-Methoxyisochinolin

Lieferant: COMBI-BLOCKS

HC-6026-5G HC-6026-1G
COBBHC-6026-5GEA 0 EUR
COBBHC-6026-5G COBBHC-6026-1G
7-Methoxyisochinolin
7-Methoxyisoquinoline
Formel: C₁₀H₉NO
MDL: MFCD09030747
CAS-Nummer: 39989-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR