2-(Ethylthio)nicotinoylchlorid

Lieferant: Thermo Scientific

BTB06382.1 BTB06382.10
MAYBBTB06382.1EA 0 EUR
MAYBBTB06382.1 MAYBBTB06382.10
2-(Ethylthio)nicotinoylchlorid
2-(Ethylthio)nicotinoylchlorid
Formel: C₈H₈ClNOS
CAS-Nummer: 123116-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR