2-(Methylthio)nicotinsäure
Lieferant: Thermo Scientific
BTB06287.10
BTB06287.50
BTB06287DA
MAYBBTB06287.10EA
0
EUR
MAYBBTB06287.10
MAYBBTB06287.50
MAYBBTB06287DA
2-(Methylthio)nicotinsäure
2-(Methylthio)nicotinsäure
Formel:
C₇H₇NO₂S Molecular Weight: 169,20 g/mol |
MDL:
MFCD00010101 CAS-Nummer: 74470-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...