2-(4-Methoxyphenoxy)-3-nitropyridin

Lieferant: BIONET RESEARCH

4Y-0710.10 4Y-0710.1 4Y-0710.500 4Y-0710.5
BIOT4Y-0710.10EA 0 EUR
BIOT4Y-0710.10 BIOT4Y-0710.1 BIOT4Y-0710.500 BIOT4Y-0710.5
2-(4-Methoxyphenoxy)-3-nitropyridin
2-(4-Methoxyphenoxy)-3-nitropyridine
Formel: C₁₂H₁₀N₂O₄
Molecular Weight: 246.2188 g/mol
MDL: MFCD00829021
CAS-Nummer: 76893-48-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR