2-(4-Methoxyphenoxy)-5-nitropyridin
Lieferant: BIONET RESEARCH
3Y-0718.1
3Y-0718.10
3Y-0718.5
3Y-0718.500
BIOT3Y-0718.1EA
0
EUR
BIOT3Y-0718.1
BIOT3Y-0718.10
BIOT3Y-0718.5
BIOT3Y-0718.500
2-(4-Methoxyphenoxy)-5-nitropyridin
2-(4-Methoxyphenoxy)-5-nitropyridine
Formel:
C₁₂H₁₀N₂O₄ |
CAS-Nummer:
131941-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...