2-(4-Chlorphenyl)-3-nitropyridin

Lieferant: BIONET RESEARCH

3Y-0713.1 3Y-0713.10 3Y-0713.500
BIOT3Y-0713.1EA 0 EUR
BIOT3Y-0713.1 BIOT3Y-0713.10 BIOT3Y-0713.500
2-(4-Chlorphenyl)-3-nitropyridin
2-(4-Chlorophenyl)-3-nitropyridine
Formel: C₁₁H₇ClN₂O₂
MDL: MFCD06200957
CAS-Nummer: 886361-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR