2-(4-Chlorphenyl)-5-nitropyridin
Lieferant: BIONET RESEARCH
3Y-0712.1
3Y-0712.10
3Y-0712.5
3Y-0712.500
BIOT3Y-0712.1EA
0
EUR
BIOT3Y-0712.1
BIOT3Y-0712.10
BIOT3Y-0712.5
BIOT3Y-0712.500
2-(4-Chlorphenyl)-5-nitropyridin
2-(4-Chlorophenyl)-5-nitropyridine
Formel:
C₁₁H₇ClN₂O₂ |
MDL:
MFCD06200956 CAS-Nummer: 874492-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...