2-Amino-3-thiophencarbonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Aminothiophene-3-carbonitrile

CFF497.5 CFF497.25 017505-5G 017505-25G
CHMPCFF497.5EA 321 EUR
CHMPCFF497.5 CHMPCFF497.25 FLUO017505-5G FLUO017505-25G
2-Amino-3-thiophencarbonitril
2-Amino-3-thiophencarbonitril
Formel: C₅H₄N₂S
Molecular Weight: 124,16 g/mol
MDL: MFCD00706298
CAS-Nummer: 4651-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR