Ethyl-2-amino-3-thiophencarboxylat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-aminothiophene-3-carboxylate

CFF182.5 CFF182.10 CFF182.25 017254-25G 017254-5G
CHMPCFF182.5EA 401 EUR
CHMPCFF182.5 CHMPCFF182.10 CHMPCFF182.25 FLUO017254-25G FLUO017254-5G
Ethyl-2-amino-3-thiophencarboxylat
Ethyl-2-amino-3-thiophencarboxylat
Formel: C₇H₉NO₂S
Molecular Weight: 171.22 g/mol
MDL: MFCD01566303
CAS-Nummer: 31891-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR