Methyl-2-amino-3-thiophencarboxylat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 2-aminothiophene-3-carboxylate

4N-801.500 4N-801.1 4N-801.5 4N-801.10
BIOT4N-801.500EA 0 EUR
BIOT4N-801.500 BIOT4N-801.1 BIOT4N-801.5 BIOT4N-801.10
Methyl-2-amino-3-thiophencarboxylat
Methyl-2-amino-3-thiophencarboxylat
Formel: C₆H₇NO₂S
Molecular Weight: 157.19 g/mol
MDL: MFCD00159547
CAS-Nummer: 4651-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR