Methyl-3-amino-2-thiophencarboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-amino-2-thiophenecarboxylate

OS-7491-100G OS-7491-25G
COBBOS-7491-100GEA 102 EUR
COBBOS-7491-100G COBBOS-7491-25G
Methyl-3-amino-2-thiophencarboxylat
Methyl-3-amino-2-thiophencarboxylat
Formel: C₆H₇NO₂S
Molecular Weight: 157.19 g/mol
MDL: MFCD00009765
CAS-Nummer: 22288-78-4
EINECS: 244-895-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR