Methyl-3-amino-2-thiophencarboxylat
Lieferant: CHEMPUR
Synonyme:
Methyl 3-amino-2-thiophenecarboxylate
FL9639.POR
009639-25G
009639-100G
CHMPFL9639.POREA
0
EUR
CHMPFL9639.POR
FLUO009639-25G
FLUO009639-100G
Methyl-3-amino-2-thiophencarboxylat
Methyl-3-amino-2-thiophencarboxylat
Formel:
C₆H₇NO₂S Molecular Weight: 157.19 g/mol |
MDL:
MFCD00009765 CAS-Nummer: 22288-78-4 EINECS: 244-895-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...