Methyl-3-amino-2-thiophencarboxylat

Lieferant: Thermo Scientific

Synonyme: Methyl 3-amino-2-thiophenecarboxylate

SB01239.1 SB01239.10 SB01239.50
MAYBSB01239.1EA 0 EUR
MAYBSB01239.1 MAYBSB01239.10 MAYBSB01239.50
Methyl-3-amino-2-thiophencarboxylat
Methyl-3-amino-2-thiophencarboxylat
Formel: C₆H₇NO₂S
Molecular Weight: 157.19 g/mol
MDL: MFCD00009765
CAS-Nummer: 22288-78-4
EINECS: 244-895-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR